Anti-HMGB3 Rabbit Polyclonal Antibody
: Avantor
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HMGB3 / HMG4, and purified by antigen affinity chromatography.
Type: Primary
Antigen: HMG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse
Frequently Bought Together