Order Entry
United States
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody [clone: 06] (FITC (Fluorescein Isothiocyanate))
Anti-CD14 Mouse Monoclonal Antibody [clone: 06] (FITC (Fluorescein Isothiocyanate))
Supplier:  Sino Biological
Certificates
undefined
Anti-CD14 Mouse Monoclonal Antibody [clone: 06] (FITC (Fluorescein Isothiocyanate))
Supplier:  Sino Biological
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 103628-748
  • 103630-418
  • Antigen Symbol:
  • CD14
  • CD14
  • Antigen Name:
  • CD14 molecule
  • CD14 molecule
  • Antigen Synonyms:
  • monocyte differentiation antigen CD14|CD14 antigen
  • monocyte differentiation antigen CD14|CD14 antigen
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 06
  • 06
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • Shipping Temperature:
  • Ambient
  • Ambient

Specifications

Related Information

Frequently Bought Together