Produced in rabbits immunized with purified, a synthetic peptide corresponding to the center region of MERS-CoV (NCoV / Novel coronavirus) Nucleocapsid Protein (NP protein), and purified by antigen affinity chromatography.
Type: Primary
Antigen: Spike Protein S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: MERS-CoV
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76778-104
- 103628-572
- Antigen Symbol:
- NP-CoV
- NP-CoV
- Antigen Name:
- Coronavirus Nucleocapsid
- Coronavirus Nucleocapsid
- Antigen Synonyms:
- Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- MERS-CoV
- MERS-CoV
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- A synthetic peptide corresponding to the center region of MERS-CoV (NCoV / Novel coronavirus) Nucleocapsid Protein (NP protein).
- A synthetic peptide corresponding to the center region of MERS-CoV (NCoV / Novel coronavirus) Nucleocapsid Protein (NP protein).
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- 0.2 μm filtered solution in PBS
- Storage Temperature:
- This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 °C. Preservative-Free. Avoid repeated freeze-thaw cycles.
- This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 °C. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:
- Ambient
- Ambient