Orders LinkContactUsLinkComponent
Xcelite® Hex Socket Type Nutdriver, Apex
Xcelite® Hex Socket Type Nutdriver, Apex
Xcelite® Hex Socket Type Nutdriver, Apex
Product Family Options
  • Description
    Blade diameter
    Blade length
    Overall length
  • Nutdriver with Chrome Finish, Hollow Shaft
    6.35 mm (¼")
    79 mm (3⅛")
    184.2 mm (7.25")
    Each
    Each - $59.98
    $59.98
    Each
       $59.98
     : THS8N
  • Nutdriver with Chrome Finish, Hollow Shaft
    5.55 mm (⁹⁄₃₂")
    79 mm (3⅛")
    184.2 mm (7.25")
    Each
    Each - $50.78
    $50.78
    Each
       $50.78
     : THS9N