- Description:Universal connector cleaner
- Size:20'
- Cat. No.:76250-014
This hand held connector cleaner is a great alternative to alcohol or solvent and wipes.
- Cleans fiber optic ferrule ends and connector bulkheads
- Dissipates static charges from connector endfaces
- Non-hazardous
The cartridge offers lint-free, clean room grade cleaning media which effectively removes dirt, dust, oil, grease and debris from connector end faces. A new resilient rubber pad beneath the cleaning surface prevents scaratching, and a sliding cover protects the cleaning media from contamination when not in use.