Weller® Ergonomic Precision with Inside-Serrated Tips, Apex Tool Group
76240-432
- Length:120 mm
- Material:Special Stainless steel/Soft Foam Handles
- Environmentally Preferable:
- Cat. No.:76240-432
Designed for standard application.
- This series offers models with thin shaped tips to suit every application
- Ergonomically shaped handles reduce hand fatigue and facilitate comfortable working
- Thermally insulated, soft foam handles
- ESD-safe
Ergonomically shaped handles reduce Carpal Tunnel Syndrome (CTS) and early hand fatigue. The product comes with two-color, thermally insulated soft grip handles made from soft foam material ensure high user comfort. Manufactured from non-magnetic, acid proof, and stainless steel alloy.