ContactUsLinkComponent
Triple-Layer Bubble Pouches, Associated Bag
Triple-Layer Bubble Pouches, Associated Bag
Catalog # 76248-010
Supplier:  Associated Bag
undefined
Triple-Layer Bubble Pouches, Associated Bag
Catalog # 76248-010
Supplier:  Associated Bag
Supplier Number:  374-4-04
Specifications
  • Description:
    Triple layer bubble pouches
  • Dimensions:
    8×11"
  • Cat. No.:
    76248-010
Specifications
About this item
Perfect for fragile items or long ship cycle applications.
  • Self-sealing fold-over closure secures contents
Triple layer construction features one layer of bubble surrounded by two layers of poly for added protection and ease of insertion.