TouchNTuff® 92-600 Gloves, Ansell
Supplier: Avantor
Certificates
About this item
TouchNTuff® 92-600 are ideal for chemical splash protection. Made with a comfortable, proprietary formulation, this durable nitrile glove offers superior splash resistance against a wide range of hazardous chemicals.
- Silicone-free design is paint and finish process friendly
- Smooth external glove surface
- Super soft nitrile provides a comfortable, silky feel for extended wear
Recommended for chemical handling, industrial production, laboratory analysis, maintenance, meat, fish and food processing, sample taking and processing, transferring liquids and solids.
Certifications: FDA 21 CFR 177-2600. Compliant with U.S. Food Contact Regulations. EN 374. CE certified.
Packaging: 100 Gloves per box; 10 boxes per carton/case.
Frequently Bought Together