ContactUsLinkComponent
Conductive Tool Boxes, Flambeau
Conductive Tool Boxes, Flambeau
Catalog # 76244-952
Supplier:  FLAMBEAU INC. TE
Conductive Tool Boxes, Flambeau
Catalog # 76244-952
Supplier:  FLAMBEAU INC. TE
Supplier Number:  14800-2C
Specifications
  • Color:
    Black
  • Description:
    Conductive Tool Boxes
  • Dimensions:
    13×53/8×41/4" (Interior)
  • Material:
    HDPE
  • Cat. No.:
    76244-952
Specifications
About this item
Conductive tool boxes with full-length tray that measures 121/4×43/4×11/2"
  • Snap-Shut latch
  • Carrying handle
  • Conductive
  • Lift-out Tray