$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Boxes
Storage Boxes
Tip Out Bins, CF Clark
Tip Out Bins, CF Clark
76247-184
:
CF CLARK ASSO. INC. TE
Tip Out Bins, CF Clark
76247-184
:
CF CLARK ASSO. INC. TE
:
6704
Description:
Tip Out Bin, 4 Bins
Dimensions:
6
3
/
4
×23
5
/
8
"
Cat. No.:
76247-184