POLY-TEMP® MD Tapes, General Purpose, Anti-Seize Technology
Supplier: ANTI-SEIZE TECHNOLOGY TE
Certificates
About this item
Thread seal tapes are made from 100% pure PTFE resins.
- Never dries out, no mess
- No dripping
- Easy to apply
Medium density allows it to be used as a general-purpose thread seal tape in most applications. Can be used on all types of pipes and most services. More economical than most paste thread sealing compounds.
Certifications: UL Listed.
Frequently Bought Together