Static Shielding Bags, 1000 Series, Z-Mar Technology
Supplier: Z-MAR TECHNOLOGY INC. TE
Certificates
About this item
These transparent metal-in static shielding bags are designed to provide a static safe environment for sensitive electronic devices.
- Heat sealable bags
- Bags are printed with an ESD protective symbol and a lot code for traceability
These bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric in concert with the metal layer provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
Frequently Bought Together