ContactUsLinkComponent
Luer Lock Syringe Barrels with Stoppers, Jensen Tools
Luer Lock Syringe Barrels with Stoppers, Jensen Tools
Catalog # 76240-626
Supplier:  JENSEN GLOBAL INC
Luer Lock Syringe Barrels with Stoppers, Jensen Tools
Catalog # 76240-626
Supplier:  JENSEN GLOBAL INC
Supplier Number:  JG30A-20
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Volume:
    30 ml
  • Color:
    Clear
  • Cat. No.:
    76240-626
Specifications
About this item
Syringe Barrels are manufactured with a heavy flange, custom designed for air dispensing and include grey plastic, drip-free stoppers.
Packaging: Conveniently packaged with barrels and stoppers in one bag.