ContactUsLinkComponent
Stencil Cleaning Roll, AGI Corporation
  76246-958
Stencil Cleaning Roll, AGI Corporation
  76246-958
 :  176216
  • Description:
    Cleaning Roll, 400 mm
  • Cat. No.:
    76246-958
This roll is a high-performance USC roll that offers excellent cleaning performance at a costeffective price.
  • High performance USC mediums
  • Developed specifically for automated under screen cleaning
  • Enhanced cleaning raises yield and throughput
  • High tear resistance raises MTBF and reduces operator intervention
  • Compatible with all DEK screen printers
The DEK SMT Roll has been developed to optimise the cleaning performance of the automated under-screen cleaning systems for DEK screen printers, and most of the MPM, EKRA, Fuji, Panasonic, Sanyo, and SMTech/Quad screen printers.