Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bags
Storage Bags
Static Bag Shields, Menda
Static Bag Shields, Menda
Static Bag Shields, Menda
Catalog # 76253-074
Supplier:
Desco
Expand image
Static Bag Shields, Menda
Catalog # 76253-074
Supplier:
Desco
Supplier Number:
ASK-36405
Specifications
Description:
Shield bag with zip top
Size:
16×24"
Cat. No.:
76253-074
Packaging:
100/pack
Specifications
About this item
Product Details
Product Details