ContactUsLinkComponent
Soder-Wick® Unfluxed Desoldering Braid, ITW Chemtronics
Soder-Wick® Unfluxed Desoldering Braid, ITW Chemtronics
Catalog # 76245-666
Supplier:  ITW Chemtronics
Soder-Wick® Unfluxed Desoldering Braid, ITW Chemtronics
Catalog # 76245-666
Supplier:  ITW Chemtronics
Supplier Number:  70-2-25
Specifications
  • Width:
    0.060"
  • Color:
    Yellow
  • Description:
    Soder-Wick® Unfluxed Desoldering Braid on Spool
  • Length:
    25'
  • Size:
    #2
  • Cat. No.:
    76245-666
Specifications
About this item
Soder-Wick® desoldering braid significantly reduces rework/repair time and minimizes the risk of heat damage to the board.
  • Can be coated with any flux type
  • Allows for a constant flux type throughout the production process
  • Provides quick and safe desoldering
The geometrically precise weave design allows for maximum capillary action and solder capacity. This unit optimize heat transfer through the braid and into the solder joint, resulting in faster wicking action than any other competitive brand.
Minimal flux residue on the board speeds up the cleaning process or eliminates it entirely.
Certifications: Meets MIL-F-14256 F, DOD-STD-883E, Method 2022, ANSI/IPC J STD-004 and NASA-STD-8739.3 Soldered Electrical Connections.
Packaging: 10' spools packaged in ESD-safe static dissipative bobbins. Sealed in nitrogen-purged packaging to avoid corrosion and loss of performance from moisture and oxygen.