Edsyn™ Soldering Tip Cleaner, Jensen Tools
76242-486
- Description:No-Wet Soldering Tip Cleaner
- Cat. No.:76242-486
Soft wire mesh design cleans tips without using water and will not reduce tip temperature as with wet sponges.
- Never needs water
Instead of using a wiping motion as with regular sponges, just thrust the iron tip straight into the mesh and remove.