Orders LinkContactUsLinkComponent
Proto® Socket Bit, Grainger
Proto® Socket Bit, Grainger
Proto® Socket Bit, Six Points, Alloy steel, 1⅞", 1/4" Hex, Full polished chrome, 5/64"
Catalog # 76253-508
Supplier:   GRAINGER
Supplier #:  426H51
Proto® Socket Bit, Grainger
Catalog # 76253-508
Supplier:  GRAINGER
Proto® Socket Bit, Grainger
Proto® Socket Bit, Six Points, Alloy steel, 1⅞", 1/4" Hex, Full polished chrome, 5/64"
Price
$45.32
Log in for availability
Each
Product Details & Documents
The Proto® socket bit has a hex opening that allows the bit holder to take the torque load.
  • Polished chrome finish
  • Insert length: 1"
Made from a heat-treated alloy for wear resistance which minimizes distortion for a long life.
Specifications
  • Description:
    Proto® Socket Bit, Six Points
  • Length:
    1⅞"
  • Material:
    Alloy steel
  • Tip size:
    5/64"
  • Finish:
    Full polished chrome
  • Drive size:
    1/4" Hex