Socket Driver Set, Testequity LLC TE
Catalog # 76242-948
Supplier: TESTEQUITY LLC TE
Specifications
- Description:Socket Driver Set
- Cat. No.:76242-948
Specifications
About this item
Delivery: This chrome plated steel sets feature a 51/2" long quick-release ratchet, a 6" long spinner handle, and 3 and 6" long extensions. Inch set includes 6-point sockets in the following sizes: 3/16, 7/32, 1/4, 9/32, 5/16, 11/32, 3/8, 7/16 and 1/2".