Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tapes
Sealing Tape, Tapecase
Sealing Tape, Tapecase
Sealing Tape, Tapecase
Catalog # 76247-526
Supplier:
TAPECASE LTD. TE
Expand image
Sealing Tape, Tapecase
Catalog # 76247-526
Supplier:
TAPECASE LTD. TE
Supplier Number:
61-1/4X36YD
Specifications
Description:
PTFE tape
Length:
½×36 yds.
Material:
PTFE
Thickness:
7 mil.
Cat. No.:
76247-526
Specifications
About this item
Product Details
Product Details