Slotted Screwdrivers, Zoro
Catalog # 76252-696
Supplier: ZORO - CUSTOMER SERVICE TE
Specifications
- Cat. No.:76252-696
- Tip size:3/8"
- Overall length:13⁷⁄₁₆"
- Finish:Chrome
- Handle:Cushion Grip
- Shank length:8"
- Tip type:Keystone slotted
Specifications
About this item
This keystone slotted screwdriver has acetate with rubber grip cushion grip handles. The square shape and steel construction make for top-grade performance.
Certifications: ASME, ANSI.