ContactUsLinkComponent
Moody® Extended Reach Precision Screwdriver Set, TESTEQUITY LLC TE
Catalog # 76242-914
Supplier:  TESTEQUITY LLC TE
Moody® Extended Reach Precision Screwdriver Set, TESTEQUITY LLC TE
Catalog # 76242-914
Supplier:  TESTEQUITY LLC TE
Supplier Number:  58-0390
Specifications
  • Description:
    Pollicis® 8-Piece Slotted/Phillips Precision Screwdriver Set
  • Cat. No.:
    76242-914
  • Overall length:
    6.3"
  • Includes:
    (5) Slotted Screwdrivers, (3) Phillips Bit Screwdrivers, (1) Vinyl Pouch
  • Drive size:
    0.078, 0.093, 0.125, 0.141, 0.156 Decimal Inch (Slotted), # 00, #0,#1 (Phillips)
Specifications
About this item
These unique, precision screwdrivers feature an ESD-Safe, ergonomic handle with a hexagonal swivel top. The blades are hardened chrome vanadium steel, and the tips have a black oxide, matte finish.