Retention Tester, DMC
Catalog # 76253-648
Supplier: DMC TE
Specifications
- Description:Hand Tools Retention Tester, 1.5 - 3.0 lbs
- Cat. No.:76253-648
- For use with:Pin and Socket Contacts
- Type:Adjustable
Specifications
About this item
Ths testers to test retention of socket contacts, use the socket tester tip with a preset tool insert tester (pin) into mating end of contact.
Tool must be in a straight line with contact. For pin contacts, fit pin tester to tool body and place over contact at the mallng end.