Polymask Masking Tapes, 3M
Supplier: 3M POLYMASK TE
Certificates
About this item
A 76 µm high-tack protective tape with a polyethylene backing.
- Provides temporary protection of many light textured or flat gloss surfaces
Non-UV applications requiring good clarity, low elongation, and good abrasion resistance.
Frequently Bought Together