Order Entry
United States
ContactUsLinkComponent
Adhesive Transfer Tape 9485PC, Tapecase
Adhesive Transfer Tape 9485PC, Tapecase
Catalog # 76242-726
Supplier:  TAPECASE LTD. TE
Adhesive Transfer Tape 9485PC, Tapecase
Catalog # 76242-726
Supplier:  TAPECASE LTD. TE
Supplier Number:  9485PC-1/4

Specifications

  • Width:
    ¼"
  • Length:
    60 yds.
  • Cat. No.:
    76242-726
  • Core size:
    76 mm (3")

Specifications

About this item

Adhesive Transfer Tape 9485PC is an excellent solution bonding high and low surface energy materials.

  • Adhesive is fiber reinforced for roll stability
  • Low odor for improved working environment
  • Excellent chemical resistance and shear and peel strength

Designed for short term temperature exposure up to 450 °F (232 °C) and long term 300 °F (149 °C). It can also be used for joining materials that have rougher surfaces.