Packing Sheets, Extra Packaging Corporation
Catalog # 76237-916
Supplier: EXTRA PACKAGING CORP. TE
Specifications
- Description:Packing Sheets, PE, Anti-static, Fire Retardant
- Thickness:6 mil.
- Cat. No.:76237-916
Specifications
About this item
Anti-static, fire retardant film that protects surfaces during periods of installation, construction, maintenance, repair, tenant improvement, hospital renovation and telecommunication work.
- Printed with FR standard and a statement
- Can be used to cover a variety of surfaces including electronic equipment
- Anti-Static nature of this film does not attract dust from its environment
A perfect protective covering for equipment sensitive to electrostatic discharge.
Certifications: Passes the NFPA 701-04 Test 1 Fire Standard and ASTM E-84 Class A.