One-Step Desoldering Braids, Easy Braid
Supplier: EASY BRAID CO TE
Certificates
About this item
No-Clean Desoldering Braids.
- Oxide-free clean copper
- Clean, fast and reliable
- More strands of copper (more surface area) per square inch
- Flux residue remaining on board does not need to be cleaned
Manufactured under SPC guidelines. High SIR conforming to Bellcore specification GR-78-CORE (TR-TSY000078), and IPC Test Method III.
To be used in conjunction with processes using RMA type no-clean fluxes, conforms to the requirements of ANSI/J-STD-004, Type LO.
Packaging: Packaged in 5, 10, 25 and 100 foot lengths.
Frequently Bought Together