Replacement Focusing Arm for ProZoom 4.5 Series Microscopes, O.C. White
76243-918
- Description:Replacement focusing arm for use with ProZoom 4.5 series microscopes
- Cat. No.:76243-918
The O.C. White SZ-FA20 Replacement Focusing Arm is designed for use with ProZoom 4.5 series microscopes.
The focusing arm includes a 20 mm hole, which is standard to the system.