Masterflex Tubing, Cole Parmer
Supplier: Avantor
Certificates
About this item
These tubing provides optimal performance / pump life hours with peristaltic pump equipment.
- Single-use custom lengths and configurations available to minimize downtime
- Excellent biocompatibility that meets multiple classifications
- Slightly greater clarity than peroxide-cured tubing
- Smooth surfaces; lower protein binding levels
- Fewer potential leachables
Autoclavable, Sterilization methods includes Ethylene Oxide, Gamma Irradiation.
Certifications: 3A;European Pharmacopoeia;USP;ADCF;FDA;Non-DEHP;RoHS.
Frequently Bought Together