Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Label and Tape Dispensers
Label Dispensers, Botron
Label Dispensers, Botron
Label Dispensers, Botron
Catalog # 76250-990
Supplier:
BOTRON COMPANY INC.
Expand image
Label Dispensers, Botron
Catalog # 76250-990
Supplier:
BOTRON COMPANY INC.
Supplier Number:
B6702
Specifications
Description:
LabeldDispenser
Cat. No.:
76250-990
Specifications
About this item
Product Details
Holds two 2" rolls or one 4" roll.
Product Details