X-ACTO® Precision Knife, Jensen Tools
76247-418
- Color:Black
- Description:Precision Knife
- Cat. No.:76247-418
- Handle length:4.63"
- Blade no.:#11
- Blade material:Carbon steel
The X-ACTO X2000 Precision Knife is the ultimate cutting tool for precision and accuracy.
- Sleek and comfortable to hold
- Built with a carbon steel blade for a reliable cut every time
- Small, detailed cuts are made with precision, accuracy and confidence
- Safety cap is included with each X2000 knife
Made from the same quality materials as the original X-ACTO knife.
The true brilliance of this precision cutting tool is in its design. You’ll be able to make even the most complex cuts with confidence and ease – no matter how small and no matter how detailed.