ContactUsLinkComponent
Hexacon Spudger Soldering Tools, Jensen Tools
Hexacon Spudger Soldering Tools, Jensen Tools
Catalog # 76244-432
Supplier:  TESTEQUITY LLC TE
Hexacon Spudger Soldering Tools, Jensen Tools
Catalog # 76244-432
Supplier:  TESTEQUITY LLC TE
Supplier Number:  MA-800G
Specifications
  • Description:
    Soldering Aid
  • Cat. No.:
    76244-432
Specifications
About this item
  • ESD safe, static-dissipative fiberglass polyamide material
  • Electrically and thermally non-conductive
  • Does not scratch printed wiring boards