ContactUsLinkComponent
Halogen Bulbs, Ushio America
Halogen Bulbs, Ushio America
Catalog # 76244-828
Halogen Bulbs, Ushio America
Catalog # 76244-828
Supplier Number:  5000506
Specifications
  • Description:
    UV Lamp
  • Environmentally Preferable:
  • Cat. No.:
    76244-828
Specifications
About this item
Used in fluorescent microscope system, spot UV curing, light guid application.
  • Enhanced electrodes designed for precise positioning and hight arc stability
  • Improved manufacturing process enable exended life