Grinding Stones, Dremel
76246-294
- Color:Green
- Material:Silicon carbide
- Size:3/16"
- Cat. No.:76246-294
- Shank diameter:0.125"
This silicon carbide grinding stone is designed to grind and etch stone, glass, ceramics, porcelain and non-ferrous metals.
- Specially designed for working with conic and flat surfaces
- Great for grinding and etching
Ideal for sharpening, deburring and general purpose grinding on harder materials.