ContactUsLinkComponent
Standard Clamp Gaskets, Newman Sanitary Gasket Company
Standard Clamp Gaskets, Newman Sanitary Gasket Company
Catalog # 76239-930
Standard Clamp Gaskets, Newman Sanitary Gasket Company
Catalog # 76239-930
Supplier Number:  40RX-PX-300
Specifications
  • Description:
    Standard Clamp Gasket, Platinum Cured, Clear, 3"
  • Material:
    Silicone
  • Cat. No.:
    76239-930
Specifications
About this item
These gaskets are used in processing pipeline systems for the food, dairy, beverage, pharmaceutical and bio tech industries.
Certifications: Meets criteria of the FDA, and accepted by the USDA, and USP Class VI certifications.