Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Gaskets
Gaskets, CPC (Colder Products Company)
Gaskets, CPC (Colder Products Company)
Gaskets, CPC (Colder Products Company)
Catalog # 76248-564
Supplier:
Colder Products
Expand image
Gaskets, CPC (Colder Products Company)
Catalog # 76248-564
Supplier:
Colder Products
Supplier Number:
621200
Specifications
Color:
Natural
Description:
Gasket
Material:
EPDM
Cat. No.:
76248-564
Specifications
About this item
Product Details
Product Details