ContactUsLinkComponent
Orifice Plate Gasket, Garlock Hygienic Technologies
  76252-636
Orifice Plate Gasket, Garlock Hygienic Technologies
  76252-636
 :  A80MPSFY-150-GAS
  • Description:
    FKM Gasket With Stainless Steel Plate
  • Material:
    FKM
  • Size:
    11/2"
  • Cat. No.:
    76252-636
Orifice plates can advance system’s performance, adjust flow rates, balance backflow and equalize backpressure during SIP procedures. Achievable benefits while maintaining sanitary conditions.