ContactUsLinkComponent
Suma® Break-Up® Heavy Duty Foaming Grease-Releaser Cleaner, Dalco
Suma® Break-Up® Heavy Duty Foaming Grease-Releaser Cleaner, Dalco
Catalog # 76241-290
Suma® Break-Up® Heavy Duty Foaming Grease-Releaser Cleaner, Dalco
Catalog # 76241-290
Supplier Number:  904495
Specifications
  • Size:
    4×3.78 L (1 gal.)
  • Cat. No.:
    76241-290
Specifications
About this item
A neutral, solvent-free degreaser for removing protein, fats grease, oils and other food soils in a food preparation environment.
  • Penetrates, lifts and dissolves soil on hard surfaces, especially where grease is a problem
  • Foam away fats, oils, grease, dirt, grime, soap scum from walls, floors and equipment
  • Cleans fast and easy without scrubbing
  • Cleans even hard to clean grout
Certifications: Kosher
NSF
Letter of No Objection