Order Entry
United States
ContactUsLinkComponent
Tritan Perma-Walk Floor Mat, Ranco Industries
Tritan Perma-Walk Floor Mat, Ranco Industries
  76249-332
Tritan Perma-Walk Floor Mat, Ranco Industries
  76249-332
 :  TPW48X6

 

  • Description:
    Tritan Perma-Walk Floor Mat
  • Length:
    122 cm (4 ft.)
  • Thickness:
    19 mm (¾")
  • Environmentally Preferable:
  • Cat. No.:
    76249-332
  • Width:
    15 cm (6")

 

 

Mat designed with one quarter inch wide flat corrugations running parallel to the length of the mat for easy sweeping and cleaning.

  • Reduces slipping and falls in heavy traffic areas, aisles, locker rooms and assembly areas

Built-in resistance to constant abuse, a characteristic of Tritan construction assures long life.