ContactUsLinkComponent
Utility File Set, Grobet File Co of America
  76242-798
 :  Grobet
Utility File Set, Grobet File Co of America
  76242-798
 :  Grobet
 :  32.51
  • Description:
    File set
  • Length:
    6½"
  • Cat. No.:
    76242-798
Set including six American Pattern file shapes: square, half-round, three-square, round, flat, and warding. File cut lengths are approximately 4" (10 cm) with an overall length of 7" (17 cm). Each file has a cushioned grip handle. Set includes a hanging pouch.