Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tool Sets
Fault Finder Locator, Fiber Instrument Sales (FIS)
Fault Finder Locator, Fiber Instrument Sales (FIS)
Fault Finder Locator, Fiber Instrument Sales (FIS)
Catalog # 76243-316
Supplier:
FIBER INSTRUMENT SALES INC. TE
Expand image
Fault Finder Locator, Fiber Instrument Sales (FIS)
Catalog # 76243-316
Supplier:
FIBER INSTRUMENT SALES INC. TE
Supplier Number:
F19000125
Specifications
Description:
Fault Finder Locator
Size:
1.25 mm
Cat. No.:
76243-316
For use with:
LC and MU Connectors
Specifications
About this item
Product Details
Product Details