ContactUsLinkComponent
Eyepiece, LX Microscopes by Unitron
Eyepiece, LX Microscopes by Unitron
Catalog # 76252-216
undefined
Eyepiece, LX Microscopes by Unitron
Catalog # 76252-216
Supplier Number:  23762
Specifications
  • Description:
    Eyepiece, 20× Magnification
  • Cat. No.:
    76252-216
Specifications
About this item
This is for use with LX Microscopes by UNITRON systems 273, 373 and VIP.
  • Super wide 23 mm field of view
  • Shipping weight 1 lbs.