- Color:Clear
- Description:Secondary Liner
- Material:Polyester
- Size:20"×180 yard
- Thickness:2.91 mil.
- Cat. No.:76250-066
Secondary liner available in a variety of constructions ranging from silicone coated densified kraft, polycoated paper and polyester liners to non-silicone polyester liners.
Release coating one or two sides by (fluoropolymer, non-silicone).