ContactUsLinkComponent
IDS™ Series Infinite Divider Boxes, Flambeau
Catalog # 76242-986
Supplier:  FLAMBEAU INC. TE
IDS™ Series Infinite Divider Boxes, Flambeau
Catalog # 76242-986
Supplier:  FLAMBEAU INC. TE
Supplier Number:  T900
Specifications
  • Color:
    Translucent/clear
  • Description:
    IDS™-Series Infinite Divider Boxes with 16 Dividers
  • Dimensions:
    131/8×87/8×115/16" (inside)
  • Material:
    PP
  • Cat. No.:
    76242-986
  • No. of compartments:
    4
Specifications
About this item
The Infinite Divider System (IDS), constructed of translucent polypropylene, is an extremely flexible organizational system.
  • 16 IDS Dividers
  • High Chemical Resistance
  • Mechanical Hinge
  • Snap-Shut Latches
  • Four Compartments
Each divider can be moved 1/16 of an inch (0.16 cm) along a ribbed compartment, allowing for customization of individual compartments.