Sea Braid, Desoldering Braid, Easy Braid
Catalog # 76253-698
Supplier: EASY BRAID CO TE
Specifications
- Width:0.075"
- Color:Green
- Description:Unfluxed Desoldering Braid
- Length:100' Spools
- Cat. No.:76253-698
Specifications
About this item
Unfluxed Desoldering Braids, ready to use with the flux of your choice.
- Oxide-free clean copper
- More strands of copper (more surface area) per square inch
- Can be used in conjunction with any flux to fit your process
- Coated to prevent oxidation
Manufactured under SPC guidelines. Cleaned and manufactured utilizing he latest technology in environmentally friendly chemicals and processes.
Available in static dissipative spools in compliance with the ESD Association Standard (Mil. Standard 2000 and ANSI/ESD S2020.
Packaging: Packaged in 5, 10, 25 and 100 foot lengths.