Order Entry
United States
ContactUsLinkComponent
Splice Clip Frame, Splicetronics Solutions
Splice Clip Frame, Splicetronics Solutions
Catalog # 76247-080
Splice Clip Frame, Splicetronics Solutions
Catalog # 76247-080
Supplier Number:  CP408

Specifications

  • Description:
    Splice Clip Frame
  • Dimensions:
    3.40×1.02"
  • Material:
    Brass
  • Thickness:
    0.004"
  • Cat. No.:
    76247-080

Specifications

About this item

Bandolier type clip/shim used with the ST410 splice tool and tape of any width.

  • Guarantees a secure connection of SMT component reels

This frame increase SMT pick and place machine output by 5 to 30%.

Packaging: 20 clips per frame.