MPT Carbon PEEK Plastic Probe and Spatula, Squared Body, ESD Safe, Ideal-tek
76251-122
: Avantor
- Description:Squared Body, Curved Fine Tip and Flat Strong Tip Chisel-Shape
- Length:150 mm
- Cat. No.:76251-122
Ideal-tek multi-purpose tools for electronics, medical device, chemistry, and watchmaking.
- Wear resistant
- Soft tips do not scratch delicate surfaces
- 150 mm length
Applications include: probe for lead-free soldering operations; positioning aid tool for assembly operations; spatula for applying adhesives and dosing chemicals in labs; stirring rod for the preparations of adhesives and solutions; scraper to remove solder masking agents, rubber latex, and adhesive coatings; use in microscopy sampling applications.