Xcelite® Transverse End Cutter, Apex
76242-744
- Description:Transverse End Cutter
- Thickness:⁹⁄₃₂"
- Cat. No.:76242-744
- Jaw width:⁷⁄₁₆"
- Jaw length:1"
- Tip size:1/8×3/16"
- Overall length:4½"
Vertical cutting in tight spaces.
Features green cushion grips.