ContactUsLinkComponent
Turrets for Crimp Tools, DMC
Turrets for Crimp Tools, DMC
Catalog # 76252-772
Supplier:  DMC TE
Turrets for Crimp Tools, DMC
Catalog # 76252-772
Supplier:  DMC TE
Supplier Number:  TP465
Specifications
  • Description:
    Single Position Head
  • Cat. No.:
    76252-772
Specifications
About this item
Single position head is used with the M22520/1-01 (AF8) tool frame and adapts the tool frame to a particular application.
The data plate on each positioner designates which contacts the positioner accommodates for its wire size and indicates selector position.
Certifications: Qualified to AS22520/1.