Order Entry
United States
ContactUsLinkComponent
Power Relay, Digi-Key
Power Relay, Digi-Key
Catalog # 76250-132
Power Relay, Digi-Key
Catalog # 76250-132
Supplier Number:  G6CU-1114P-US-

Specifications

  • Description:
    General Purpose Relay SPST-NO (1 Form A)
  • Operating Temperature:
    −25...+70 °C
  • Cat. No.:
    76250-132

Specifications

About this item

This general purpose coil through hole power relay is designed with cadmium free, silver alloy contact material.

  • Operate time: 10 ms
  • Coil resistance: 125 Ω

Coil voltage and current is measured as 5 V and 40 mA respectively.